Assay Details
| Gene Symbol: EEF2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013302.3 | NP_037434.1 | 2 - 3 | 733 | 144 | |
| XM_017023193.1 | XP_016878682.1 | 2 - 3 | 444 | 144 | ||
| XM_017023194.1 | XP_016878683.1 | 2 - 3 | 740 | 144 | ||
| GenBank mRNA | AK308757.1 | - | 2 - 3 | 678 | 144 | |
| BC032665.1 | - | 3 - 4 | 444 | 5553055 | 144 | |
| U93850.1 | - | 1 - 2 | 248 | 144 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000243.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to ischemia
translational elongation
regulation of protein autophosphorylation
cellular response to insulin stimulus
negative regulation of apoptotic process
positive regulation of endocytosis
protein autophosphorylation
positive regulation of synapse assembly
positive regulation of dendritic spine morphogenesis
cellular response to calcium ion
cellular response to cAMP
cellular response to anoxia
cellular response to brain-derived neurotrophic factor stimulus
response to prolactin
protein kinase activity
elongation factor-2 kinase activity
calcium ion binding
calmodulin binding
ATP binding
translation factor activity, RNA binding
response to ischemia
translational elongation
regulation of protein autophosphorylation
cellular response to insulin stimulus
negative regulation of apoptotic process
positive regulation of endocytosis
protein autophosphorylation
positive regulation of synapse assembly
positive regulation of dendritic spine morphogenesis
cellular response to calcium ion
cellular response to cAMP
cellular response to anoxia
cellular response to brain-derived neurotrophic factor stimulus
response to prolactin
protein kinase activity
elongation factor-2 kinase activity
calcium ion binding
calmodulin binding
ATP binding
translation factor activity, RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000561791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000561791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000561791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16003562.hg.1 | ENST00000263026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16003562.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16003562.hg.1 | TC16000243.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600151559.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600151559.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600151559.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600151559.hg.1 | TC1600007191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600151559.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600151559.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600151559.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000263026.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000561791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000561791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000561791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000563555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | ENST00000568269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007191.hg.1 | NM_013302.3 | Refseq |

EEF2K
Genomic Map