Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033438.3 | NP_254273.2 | 3 - 4 | 71 | ||
| XM_017002756.1 | XP_016858245.1 | 4 - 5 | 71 | |||
| GenBank mRNA | AF275725.1 | - | 3 - 4 | 71 | ||
| AY034613.1 | - | 3 - 4 | 71 | |||
| BC074754.2 | - | 3 - 4 | 30915213 | 71 | ||
| CB528634.1 | - | 1 - 2 | 396 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003394.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053005.hg.1 | ENST00000368092.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053005.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053005.hg.1 | ENST00000466773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053005.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053005.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053005.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053005.hg.1 | TC01003394.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053006.hg.1 | ENST00000368092.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053006.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053006.hg.1 | ENST00000466773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053006.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053006.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053006.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053006.hg.1 | TC01003394.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222391.hg.1 | TC0100016115.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222393.hg.1 | TC0100016115.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100222392.hg.1 | TC0100016115.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368092.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000368093.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000466773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | ENST00000489098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_001146173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016115.hg.1 | NM_033438.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 177405 | - | - |

SLAMF9
Genomic Map