Assay Details
| Gene Symbol: STAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015136.2 | NP_055951.2 | 14 - 15 | 1656 | 67 | |
| XM_005264973.3 | XP_005265030.1 | 14 - 15 | 1658 | 67 | ||
| XM_005264974.2 | XP_005265031.1 | 14 - 15 | 1658 | 67 | ||
| XM_006713065.1 | XP_006713128.1 | 14 - 15 | 1658 | 67 | ||
| XM_017005998.1 | XP_016861487.1 | 14 - 15 | 1658 | 67 | ||
| XM_017005999.1 | XP_016861488.1 | 14 - 15 | 1658 | 67 | ||
| XM_017006000.1 | XP_016861489.1 | 14 - 15 | 1658 | 67 | ||
| XM_017006001.1 | XP_016861490.1 | 14 - 15 | 1658 | 67 | ||
| XM_017006002.1 | XP_016861491.1 | 14 - 15 | 1658 | 67 | ||
| XM_017006003.1 | XP_016861492.1 | 14 - 15 | 1658 | 67 | ||
| XM_017006004.1 | XP_016861493.1 | 14 - 15 | 1658 | 67 | ||
| GenBank mRNA | AB052956.1 | - | 14 - 15 | 1643 | 67 | |
| AB052957.1 | - | 14 - 15 | 1637 | 67 | ||
| AJ275213.1 | - | 14 - 15 | 1612 | 67 | ||
| BC150250.1 | - | 14 - 15 | 1662 | 9007475 | 67 | |
| D87433.2 | - | 14 - 15 | 1641 | 67 | ||
| DQ786774.1 | - | 14 - 15 | 1606 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000336.hg.1
Set Membership: |
|

STAB1
Genomic Map