Assay Details
| Gene Symbol: LPAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017027468.1 | XP_016882957.1 | 1 - 2 | 107 | 93 | |
| GenBank mRNA | AK123043.1 | - | 2 - 3 | 576 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK123043.1, XM_017027468.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000589311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000589311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000589311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000589311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000592061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000592061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000592061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | ENST00000592061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000407877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000542587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000586703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000588461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000589311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000589311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000589311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000589311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000590629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000591042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000592061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000592061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000592061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | ENST00000592061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | NM_004720.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010073.hg.1 | NM_004720.5 | Refseq |

LPAR2
Genomic Map