| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009887.hg.1 |
ENST00000218104.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009887.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009887.hg.1 |
TC0X000755.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000370129.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000370129.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000370129.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009886.hg.1 |
ENST00000218104.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009886.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009886.hg.1 |
TC0X000755.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00159928.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159928.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159928.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159928.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159928.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159928.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159928.hg.1 |
TC0X00008795.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000370129.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000370129.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000370129.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008795.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159929.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159929.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159929.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159929.hg.1 |
ENST00000218104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159929.hg.1 |
ENST00000443684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159929.hg.1 |
NM_000033.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159929.hg.1 |
TC0X00008795.hg.1 |
Clariom D TC |