Assay Details
| Gene Symbol: GPR19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011520623.2 | XP_011518925.1 | 3 - 4 | 808 | 142 | |
| XM_011520624.2 | XP_011518926.1 | 3 - 4 | 462 | 142 | ||
| XM_017019212.1 | XP_016874701.1 | 3 - 4 | 1320 | 142 | ||
| XM_017019213.1 | XP_016874702.1 | 3 - 4 | 1516 | 142 | ||
| XM_017019214.1 | XP_016874703.1 | 4 - 5 | 1673 | 142 | ||
| XM_017019215.1 | XP_016874704.1 | 1 - 2 | 2650 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011520623.2, XM_011520624.2, XM_017019212.1, XM_017019213.1, XM_017019214.1, XM_017019215.1 transcripts.
Set Membership: |
|

GPR19
Genomic Map