Assay Details
| Gene Symbol: ELK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001973.3 | NP_001964.2 | 4 - 5 | 2448 | 61 | |
| XM_005244950.4 | XP_005245007.1 | 5 - 6 | 1358 | 61 | ||
| XM_005244951.4 | XP_005245008.1 | 4 - 5 | 2152 | 61 | ||
| GenBank mRNA | BC040145.1 | - | 4 - 5 | 1469 | 61 | |
| M85165.1 | - | 4 - 5 | 1347 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003750.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell differentiation
histone H3 deacetylation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
protein binding
sequence-specific DNA binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell differentiation
histone H3 deacetylation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
protein binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01057500.hg.1 | ENST00000357992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057500.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057500.hg.1 | TC01003750.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000468523.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000468523.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000468523.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000617725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000617725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000617725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | ENST00000617725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057501.hg.1 | ENST00000357992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057501.hg.1 | ENST00000539916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057501.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057501.hg.1 | TC01003750.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000289703.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000468523.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000468523.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000468523.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000617725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000617725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000617725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | ENST00000617725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017072.hg.1 | NM_021795.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229341.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229341.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229341.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229341.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229341.hg.1 | TC0100017072.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229343.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229343.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229343.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229343.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229343.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229343.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229343.hg.1 | TC0100017072.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229344.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229344.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229344.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229344.hg.1 | ENST00000357992.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229344.hg.1 | ENST00000616704.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229344.hg.1 | NM_001973.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229344.hg.1 | TC0100017072.hg.1 | Clariom D TC |

ELK4
Genomic Map