Assay Details
| Gene Symbol: AP1AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018569.5 | NP_061039.3 | 5 - 6 | 640 | 113 | |
| NR_136622.1 | 5 - 6 | 640 | 113 | |||
| XM_017008378.1 | XP_016863867.1 | 6 - 7 | 799 | 113 | ||
| XM_017008380.1 | XP_016863869.1 | 5 - 6 | 323 | 113 | ||
| GenBank mRNA | AK310787.1 | - | 5 - 6 | 639 | 113 | |
| AK315299.1 | - | 5 - 6 | 540 | 113 | ||
| AL136628.1 | - | 5 - 6 | 558 | 113 | ||
| BM823928.1 | - | 5 - 6 | 252 | 113 | ||
| BX647702.1 | - | 5 - 6 | 593 | 113 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000579.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017008380.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of receptor recycling
protein transport
regulation of Arp2/3 complex-mediated actin nucleation
cellular protein localization
vesicle targeting, trans-Golgi to endosome
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of cell motility
kinesin binding
AP-1 adaptor complex binding
negative regulation of receptor recycling
protein transport
regulation of Arp2/3 complex-mediated actin nucleation
cellular protein localization
vesicle targeting, trans-Golgi to endosome
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of cell motility
kinesin binding
AP-1 adaptor complex binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000506522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000506522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000506522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000506522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000512741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000512741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | ENST00000512741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008393.hg.1 | ENST00000274000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008393.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008393.hg.1 | TC04000579.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400157906.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400157906.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400157906.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400157906.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400157906.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400157906.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400157906.hg.1 | TC0400008427.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000274000.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000309703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000502954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000506522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000506522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000506522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000506522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000510527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000512741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000512741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | ENST00000512741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_001128426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008427.hg.1 | NM_018569.5 | Refseq |

AP1AR
Genomic Map