Assay Details
| Gene Symbol: RGS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204417.2 | NP_001191346.1 | 16 - 17 | 1654 | 120 | |
| NM_001204421.2 | NP_001191350.1 | 15 - 16 | 1543 | 120 | ||
| XM_017021834.1 | XP_016877323.1 | 14 - 15 | 1209 | 120 | ||
| GenBank mRNA | AF073921.1 | - | 11 - 12 | 1017 | 120 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000435.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
protein binding
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000343854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000404301.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000406236.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000407322.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000434263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000535521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000553525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000554782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | ENST00000556437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | NM_001204416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | NM_001204417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | NM_001204419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | NM_001204421.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | NM_001204424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005264.hg.1 | TC14000435.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | ENST00000553530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | ENST00000554782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204418.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204421.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_001204424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | NM_004296.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005279.hg.1 | TC14000435.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000343854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000402788.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000553530.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000554300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000554474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000554782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000622468.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204417.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204418.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204421.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_004296.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000343854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000402788.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000553530.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000554300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000554474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000554782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | ENST00000622468.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204417.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204418.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204421.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_001204424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | NM_004296.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153458.hg.1 | TC1400007603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000343854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000404301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000406236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000407322.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000434263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000553525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000554300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000554474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000554782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000556437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000622468.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204417.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204421.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000343854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000404301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000406236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000407322.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000434263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000553525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000554300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000554474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000554782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000556437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | ENST00000622468.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204417.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204421.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | NM_001204424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153445.hg.1 | TC1400007603.hg.1 | Clariom D TC |

RGS6
Genomic Map