Assay Details
| Gene Symbol: PPP1R8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002713.3 | NP_002704.1 | 3 - 4 | 770 | 76 | |
| XM_017001594.1 | XP_016857083.1 | 3 - 4 | 290 | 76 | ||
| GenBank mRNA | AK297771.1 | - | 3 - 4 | 296 | 76 | |
| AK313814.1 | - | 3 - 4 | 276 | 76 | ||
| U14575.1 | - | 3 - 4 | 757 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000370.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297771.1,AK313814.1,NM_002713.3,U14575.1,XM_017001594.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA catabolic process
cell proliferation
RNA splicing
negative regulation of catalytic activity
RNA phosphodiester bond hydrolysis
DNA binding
RNA binding
endonuclease activity
protein serine/threonine phosphatase inhibitor activity
protein binding
protein phosphatase type 1 regulator activity
ribonuclease E activity
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA catabolic process
cell proliferation
RNA splicing
negative regulation of catalytic activity
RNA phosphodiester bond hydrolysis
DNA binding
RNA binding
endonuclease activity
protein serine/threonine phosphatase inhibitor activity
protein binding
protein phosphatase type 1 regulator activity
ribonuclease E activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01005710.hg.1 | ENST00000236412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005710.hg.1 | ENST00000311772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005710.hg.1 | ENST00000373931.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005710.hg.1 | ENST00000399118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005710.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005710.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005710.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005710.hg.1 | TC01000370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | ENST00000236412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | ENST00000311772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | ENST00000373931.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | ENST00000399118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | ENST00000434313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01005709.hg.1 | TC01000370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000486634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | ENST00000486634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000486634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | ENST00000486634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007538.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | ENST00000236412.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | ENST00000311772.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | ENST00000373931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | ENST00000431586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | NM_002713.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | NM_014110.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | NM_138558.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100152718.hg.1 | TC0100007538.hg.1 | Clariom D TC |

PPP1R8
Genomic Map