| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031203.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031203.hg.1 |
ENST00000539153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031203.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031203.hg.1 |
TC01002049.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000462488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000462488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000462488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
ENST00000462488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000355360.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000462488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000462488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000462488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
ENST00000462488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_001017434.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012379.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100190723.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100190723.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100190723.hg.1 |
ENST00000329291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100190723.hg.1 |
NM_170725.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100190723.hg.1 |
TC0100012379.hg.1 |
Clariom D TC |