| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012211.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012211.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012211.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012211.hg.1 |
TC08000960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012209.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012209.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012209.hg.1 |
TC08000960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012212.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012212.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012212.hg.1 |
TC08000960.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800165197.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165197.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165197.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165197.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165197.hg.1 |
TC0800009569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800165193.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165193.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165193.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165193.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165193.hg.1 |
TC0800009569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800165195.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165195.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165195.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165195.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165195.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165195.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165195.hg.1 |
TC0800009569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
ENST00000329335.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
ENST00000382483.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009569.hg.1 |
NM_178857.5 |
Refseq |