Assay Details
| Gene Symbol: SLC13A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324400.1 | NP_001311329.1 | 14 - 15 | 1771 | 110 | |
| NM_022444.3 | NP_071889.2 | 11 - 12 | 1270 | 110 | ||
| XM_011516515.2 | XP_011514817.1 | 12 - 13 | 1442 | 110 | ||
| XM_011516516.2 | XP_011514818.1 | 11 - 12 | 1340 | 110 | ||
| XM_011516517.2 | XP_011514819.1 | 13 - 14 | 1561 | 110 | ||
| XM_017012554.1 | XP_016868043.1 | 11 - 12 | 1328 | 110 | ||
| GenBank mRNA | AF260824.1 | - | 11 - 12 | 1270 | 110 | |
| AK026413.1 | - | 11 - 12 | 1268 | 110 | ||
| AK314537.1 | - | 11 - 12 | 1268 | 110 | ||
| BC111775.1 | - | 14 - 15 | 1771 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001818.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07028158.hg.1 | ENST00000194130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028158.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028158.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028158.hg.1 | TC07001818.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012453.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | NM_022444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | ENST00000194130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | ENST00000427975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | ENST00000439260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | ENST00000539873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189696.hg.1 | TC0700012453.hg.1 | Clariom D TC |

SLC13A1
Genomic Map