Assay Details
| Gene Symbol: CDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317224.1 | NP_001304153.1 | 6 - 7 | 1265 | 70 | |
| NM_006727.4 | NP_006718.2 | 7 - 8 | 1764 | 70 | ||
| XM_011513923.2 | XP_011512225.1 | 8 - 9 | 1803 | 70 | ||
| XM_017008915.1 | XP_016864404.1 | 7 - 8 | 1722 | 70 | ||
| GenBank mRNA | AB035303.1 | - | 7 - 8 | 1588 | 70 | |
| KJ534806.1 | - | 6 - 7 | 1256 | 70 | ||
| KJ534808.1 | - | 6 - 7 | 1256 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001205.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05017112.hg.1 | ENST00000264463.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017112.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017112.hg.1 | TC05001205.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171889.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171889.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171889.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171889.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171889.hg.1 | TC0500010308.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000264463.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000502921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000503958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | ENST00000510477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010308.hg.1 | NM_006727.4 | Refseq |

CDH10
Genomic Map