Assay Details
| Gene Symbol: ITGB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002214.2 | NP_002205.1 | 10 - 11 | 2395 | 140 | |
| XM_011515393.2 | XP_011513695.1 | 11 - 12 | 2198 | 140 | ||
| XM_011515394.2 | XP_011513696.1 | 11 - 12 | 1796 | 140 | ||
| XM_011515396.1 | XP_011513698.1 | 11 - 12 | 2467 | 140 | ||
| XM_017012178.1 | XP_016867667.1 | 11 - 12 | 2092 | 140 | ||
| XM_017012179.1 | XP_016867668.1 | 11 - 12 | 2551 | 140 | ||
| XM_017012180.1 | XP_016867669.1 | 11 - 12 | 2844 | 140 | ||
| XM_017012181.1 | XP_016867670.1 | 12 - 13 | 2663 | 140 | ||
| XM_017012182.1 | XP_016867671.1 | 11 - 12 | 1778 | 140 | ||
| XM_017012183.1 | XP_016867672.1 | 11 - 12 | 1735 | 140 | ||
| GenBank mRNA | AK295044.1 | - | 11 - 12 | 2467 | 140 | |
| M73780.1 | - | 10 - 11 | 2370 | 140 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000125.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside metabolic process
cell adhesion
cell-matrix adhesion
integrin-mediated signaling pathway
positive regulation of gene expression
negative regulation of gene expression
extracellular matrix organization
cartilage development
placenta blood vessel development
receptor activity
receptor binding
extracellular matrix protein binding
ganglioside metabolic process
cell adhesion
cell-matrix adhesion
integrin-mediated signaling pathway
positive regulation of gene expression
negative regulation of gene expression
extracellular matrix organization
cartilage development
placenta blood vessel development
receptor activity
receptor binding
extracellular matrix protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07001923.hg.1 | ENST00000222573.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07001923.hg.1 | ENST00000537992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07001923.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07001923.hg.1 | TC07000125.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000460204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000460204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000460204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000460204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700147301.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147301.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147301.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700147301.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147301.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700147301.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700147301.hg.1 | TC0700006844.hg.1 | Clariom D TC |

ITGB8
Genomic Map