Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138348.4 | NP_612357.4 | 2 - 3 | 378 | 92 | |
| XM_011514151.2 | XP_011512453.1 | 2 - 3 | 4258 | 92 | ||
| XM_011514152.2 | XP_011512454.1 | 2 - 3 | 4258 | 92 | ||
| XM_011514154.2 | XP_011512456.1 | 2 - 3 | 4258 | 92 | ||
| GenBank mRNA | BC015392.1 | - | 2 - 3 | 304 | 4430188 | 92 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000086.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of NF-kappaB transcription factor activity
innate immune response
negative regulation of inflammatory response
canonical Wnt signaling pathway
nucleotide-binding oligomerization domain containing 2 signaling pathway
protein linear deubiquitination
thiol-dependent ubiquitin-specific protease activity
protein binding
cysteine-type peptidase activity
sprouting angiogenesis
regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of NF-kappaB transcription factor activity
innate immune response
negative regulation of inflammatory response
canonical Wnt signaling pathway
nucleotide-binding oligomerization domain containing 2 signaling pathway
protein linear deubiquitination
thiol-dependent ubiquitin-specific protease activity
protein binding
cysteine-type peptidase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05001039.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001039.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001039.hg.1 | TC05000086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001040.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001040.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001040.hg.1 | TC05000086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001043.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001043.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001043.hg.1 | TC05000086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001042.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001042.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001042.hg.1 | TC05000086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001037.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001037.hg.1 | TC05000086.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500146514.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146514.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146514.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146514.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146514.hg.1 | TC0500006822.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146523.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146523.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146523.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146523.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146523.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146523.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146523.hg.1 | TC0500006822.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146516.hg.1 | TC0500006822.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146517.hg.1 | TC0500006822.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500146524.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146524.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146524.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146524.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146524.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146524.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146524.hg.1 | TC0500006822.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500146519.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146519.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146519.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146519.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146519.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500146519.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500146519.hg.1 | TC0500006822.hg.1 | Clariom D TC |

OTULIN
Genomic Map