Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002153.2 | NP_002144.1 | 1 - 2 | 441 | 91 | |
| GenBank mRNA | AK313109.1 | - | 1 - 2 | 416 | 91 | |
| AU139034.1 | - | 1 - 2 | 416 | 91 | ||
| BC009581.1 | - | 1 - 2 | 354 | 4077164 | 91 | |
| BC059170.1 | - | 1 - 2 | 354 | 4079440 | 91 | |
| BP270993.1 | - | 1 - 2 | 462 | 91 | ||
| L11708.1 | - | 1 - 2 | 352 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000645.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
placenta development
steroid biosynthetic process
response to retinoic acid
oxidation-reduction process
estradiol 17-beta-dehydrogenase activity
17-alpha,20-alpha-dihydroxypregn-4-en-3-one dehydrogenase activity
testosterone dehydrogenase (NAD+) activity
in utero embryonic development
placenta development
steroid biosynthetic process
response to retinoic acid
oxidation-reduction process
estradiol 17-beta-dehydrogenase activity
17-alpha,20-alpha-dihydroxypregn-4-en-3-one dehydrogenase activity
testosterone dehydrogenase (NAD+) activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16009114.hg.1 | ENST00000199936.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009114.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009114.hg.1 | TC16000645.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009115.hg.1 | ENST00000199936.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009115.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009115.hg.1 | TC16000645.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009113.hg.1 | ENST00000199936.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009113.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009113.hg.1 | TC16000645.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | TC1600008609.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164245.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164242.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164242.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164242.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164242.hg.1 | TC1600008609.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164242.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164242.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164242.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164244.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164244.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164244.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164244.hg.1 | TC1600008609.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164244.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164244.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164244.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164243.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164243.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164243.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164243.hg.1 | TC1600008609.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164243.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164243.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164243.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | TC1600008609.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164248.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000566838.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008609.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | NM_002153.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | TC1600008609.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | ENST00000199936.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | ENST00000563491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | ENST00000568090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | ENST00000569351.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164247.hg.1 | NM_002153.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 195707 | - | - |

HSD17B2
Genomic Map