Assay Details
| Gene Symbol: SLC27A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003645.3 | NP_003636.2 | 3 - 4 | 1082 | 81 | |
| GenBank mRNA | AF096290.1 | - | 3 - 4 | 1054 | 81 | |
| AK123905.1 | - | 4 - 5 | 724 | 81 | ||
| AK222859.1 | - | 3 - 4 | 1053 | 81 | ||
| AK223145.1 | - | 3 - 4 | 1051 | 81 | ||
| AK290262.1 | - | 3 - 4 | 1082 | 81 | ||
| D88308.1 | - | 3 - 4 | 1072 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000391.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid alpha-oxidation
long-chain fatty acid metabolic process
fatty acid beta-oxidation
bile acid biosynthetic process
fatty acid transport
very long-chain fatty acid catabolic process
long-chain fatty acid import
methyl-branched fatty acid metabolic process
long-chain fatty acid-CoA ligase activity
receptor binding
ATP binding
fatty acid transporter activity
enzyme binding
very long-chain fatty acid-CoA ligase activity
phytanate-CoA ligase activity
pristanate-CoA ligase activity
decanoate--CoA ligase activity
fatty acid alpha-oxidation
long-chain fatty acid metabolic process
fatty acid beta-oxidation
bile acid biosynthetic process
fatty acid transport
very long-chain fatty acid catabolic process
long-chain fatty acid import
methyl-branched fatty acid metabolic process
long-chain fatty acid-CoA ligase activity
receptor binding
ATP binding
fatty acid transporter activity
enzyme binding
very long-chain fatty acid-CoA ligase activity
phytanate-CoA ligase activity
pristanate-CoA ligase activity
decanoate--CoA ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15003643.hg.1 | ENST00000267842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003643.hg.1 | ENST00000380902.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003643.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003643.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003643.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003643.hg.1 | TC15000391.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | ENST00000267842.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | ENST00000380902.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | ENST00000544960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | NM_001159629.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | NM_003645.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151307.hg.1 | TC1500007196.hg.1 | Clariom D TC |

SLC27A2
Genomic Map