Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003648.hg.1 |
ENST00000267842.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003648.hg.1 |
ENST00000380902.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003648.hg.1 |
ENST00000544960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003648.hg.1 |
NM_001159629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003648.hg.1 |
NM_003645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003648.hg.1 |
TC15000391.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
ENST00000267842.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
ENST00000380902.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
ENST00000544960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
NM_001159629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
NM_003645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
ENST00000267842.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
ENST00000380902.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
ENST00000544960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
NM_001159629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
NM_003645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151314.hg.1 |
TC1500007196.hg.1 |
Clariom D TC |