Assay Details
| Gene Symbol: CPXM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198148.2 | NP_937791.2 | 1 - 2 | 630 | 100 | |
| XM_005269528.3 | XP_005269585.1 | 1 - 2 | 480 | 100 | ||
| XM_011539283.2 | XP_011537585.1 | 1 - 2 | 480 | 100 | ||
| GenBank mRNA | AK304023.1 | - | 1 - 2 | 404 | 100 | |
| AK304814.1 | - | 1 - 2 | 441 | 100 | ||
| AY358565.1 | - | 1 - 2 | 630 | 100 | ||
| BC036789.1 | - | 1 - 2 | 460 | 5752707 | 100 | |
| BC137492.1 | - | 1 - 2 | 625 | 9021492 | 100 | |
| BC146862.1 | - | 1 - 2 | 625 | 9056790 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001721.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10022837.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022837.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022837.hg.1 | TC10001721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022838.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022838.hg.1 | TC10001721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022833.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022833.hg.1 | ENST00000368854.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022833.hg.1 | ENST00000418782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022833.hg.1 | ENST00000474277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022833.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022833.hg.1 | TC10001721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000481775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000481775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022836.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022836.hg.1 | ENST00000418782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022836.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022836.hg.1 | TC10001721.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000182825.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182825.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182825.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182825.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182825.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182825.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182825.hg.1 | TC1000012108.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000481775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000481775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | ENST00000481775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | ENST00000368854.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | ENST00000481775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182823.hg.1 | TC1000012108.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000182827.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182827.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182827.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182827.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182827.hg.1 | TC1000012108.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000182826.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182826.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182826.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182826.hg.1 | ENST00000241305.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182826.hg.1 | ENST00000615851.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182826.hg.1 | NM_198148.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182826.hg.1 | TC1000012108.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 139315 | - | - |

CPXM2
Genomic Map