| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022821.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022821.hg.1 |
ENST00000368854.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022821.hg.1 |
ENST00000418782.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022821.hg.1 |
ENST00000474277.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022821.hg.1 |
ENST00000540123.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022821.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022821.hg.1 |
TC10001721.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000481775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000481775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022820.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022820.hg.1 |
ENST00000368854.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022820.hg.1 |
ENST00000474277.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022820.hg.1 |
ENST00000540123.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022820.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022820.hg.1 |
TC10001721.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000182809.hg.1 |
TC1000012108.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000481775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000481775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |