| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017103.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017103.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017103.hg.1 |
TC04001258.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000510856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000510856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
ENST00000510856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017105.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017105.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017105.hg.1 |
TC04001258.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000510856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000510856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
ENST00000510856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010901.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174153.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174153.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174153.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174153.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174153.hg.1 |
ENST00000502365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174153.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174153.hg.1 |
TC0400010901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174151.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174151.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174151.hg.1 |
ENST00000332644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174151.hg.1 |
NM_182502.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174151.hg.1 |
TC0400010901.hg.1 |
Clariom D TC |