Assay Details
| Gene Symbol: FIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014845.5 | NP_055660.1 | 3 - 4 | 502 | 98 | |
| XM_011536281.2 | XP_011534583.1 | 3 - 4 | 1034 | 98 | ||
| XM_017011591.1 | XP_016867080.1 | 3 - 4 | 507 | 98 | ||
| XM_017011593.1 | XP_016867082.1 | 3 - 4 | 435 | 98 | ||
| GenBank mRNA | AK222732.1 | - | 3 - 4 | 399 | 98 | |
| AK291671.1 | - | 3 - 4 | 427 | 98 | ||
| BC041338.2 | - | 3 - 4 | 458 | 5274004 | 98 | |
| D87464.1 | - | 3 - 4 | 411 | 98 | ||
| DA811740.1 | - | 3 - 4 | 507 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000879.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017011593.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
vacuole organization
locomotory behavior
positive regulation of neuron projection development
dephosphorylation
negative regulation of myelination
myelin assembly
phosphatidylinositol-3-phosphate biosynthetic process
pigmentation
neuron development
phosphatidylinositol-3-phosphatase activity
protein binding
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
phosphatidylinositol biosynthetic process
vacuole organization
locomotory behavior
positive regulation of neuron projection development
dephosphorylation
negative regulation of myelination
myelin assembly
phosphatidylinositol-3-phosphate biosynthetic process
pigmentation
neuron development
phosphatidylinositol-3-phosphatase activity
protein binding
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06010360.hg.1 | ENST00000230124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010360.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010360.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010360.hg.1 | TC06000879.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010361.hg.1 | ENST00000230124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010361.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010361.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010361.hg.1 | TC06000879.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162538.hg.1 | TC0600009102.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162540.hg.1 | TC0600009102.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |

FIG4
Genomic Map