Assay Details
| Gene Symbol: FIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014845.5 | NP_055660.1 | 8 - 9 | 1086 | 68 | |
| XM_011536281.2 | XP_011534583.1 | 8 - 9 | 1618 | 68 | ||
| XM_017011591.1 | XP_016867080.1 | 8 - 9 | 1091 | 68 | ||
| XM_017011592.1 | XP_016867081.1 | 7 - 8 | 854 | 68 | ||
| GenBank mRNA | AK222732.1 | - | 8 - 9 | 983 | 68 | |
| BC041338.2 | - | 8 - 9 | 1042 | 5274004 | 68 | |
| D87464.1 | - | 8 - 9 | 995 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000879.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
vacuole organization
locomotory behavior
positive regulation of neuron projection development
dephosphorylation
negative regulation of myelination
myelin assembly
phosphatidylinositol-3-phosphate biosynthetic process
pigmentation
neuron development
phosphatidylinositol-3-phosphatase activity
protein binding
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
phosphatidylinositol biosynthetic process
vacuole organization
locomotory behavior
positive regulation of neuron projection development
dephosphorylation
negative regulation of myelination
myelin assembly
phosphatidylinositol-3-phosphate biosynthetic process
pigmentation
neuron development
phosphatidylinositol-3-phosphatase activity
protein binding
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06010366.hg.1 | ENST00000230124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010366.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010366.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010366.hg.1 | TC06000879.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162550.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162550.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162550.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162550.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162550.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162550.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162550.hg.1 | TC0600009102.hg.1 | Clariom D TC |

FIG4
Genomic Map