| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010515.hg.1 |
ENST00000306730.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010515.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010515.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010515.hg.1 |
TC15001189.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010518.hg.1 |
ENST00000306730.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010518.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010518.hg.1 |
TC15001189.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010519.hg.1 |
ENST00000306730.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010519.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15010519.hg.1 |
TC15001189.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500164723.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164723.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500164723.hg.1 |
TC1500008971.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500164723.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164723.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500164722.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164722.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500164722.hg.1 |
TC1500008971.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500164722.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164722.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
ENST00000560649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
TC1500008971.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
ENST00000558136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
ENST00000560649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164718.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500164724.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164724.hg.1 |
NM_020371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500164724.hg.1 |
TC1500008971.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500164724.hg.1 |
ENST00000306730.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500164724.hg.1 |
NM_020371.2 |
Refseq |