Assay Details
| Gene Symbol: NHSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020464.1 | NP_065197.1 | 4 - 5 | 1297 | 63 | |
| XM_011535968.2 | XP_011534270.1 | 5 - 6 | 3003 | 63 | ||
| GenBank mRNA | BX647246.1 | - | 1 - 2 | 190 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002159.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027200.hg.1 | ENST00000343505.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027200.hg.1 | ENST00000427025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027200.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027200.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027200.hg.1 | TC06002159.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027201.hg.1 | ENST00000343505.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027201.hg.1 | ENST00000427025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027201.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027201.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027201.hg.1 | TC06002159.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202705.hg.1 | TC0600014348.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202706.hg.1 | TC0600014348.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 210061 | - | - |

NHSL1
Genomic Map