Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC497048 | KU-MEL-3 | -197.7kb | Anti-sense | Chr. 6 - 4610565 - 4611919 on NCBI Build GRCh38 | |
| Downstream | ribonuclease P/MRP subunit p40 | RPP40 |
RNASEP1, bA428J1.3 |
181.0kb | Sense | Chr. 6 - 4992405 - 5004063 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map