Assay Details
| Gene Symbol: GLMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319683.1 | NP_001306612.1 | 4 - 5 | 399 | 129 | |
| NM_053274.2 | NP_444504.1 | 4 - 5 | 399 | 129 | ||
| NR_135089.1 | 4 - 5 | 399 | 129 | |||
| XM_005270401.3 | XP_005270458.1 | 3 - 4 | 261 | 129 | ||
| XM_011540546.2 | XP_011538848.1 | 6 - 7 | 562 | 129 | ||
| XM_017000137.1 | XP_016855626.1 | 4 - 5 | 767 | 129 | ||
| XM_017000138.1 | XP_016855627.1 | 4 - 5 | 767 | 129 | ||
| XM_017000139.1 | XP_016855628.1 | 4 - 5 | 767 | 129 | ||
| XM_017000140.1 | XP_016855629.1 | 3 - 4 | 377 | 129 | ||
| XM_017000141.1 | XP_016855630.1 | 4 - 5 | 399 | 129 | ||
| GenBank mRNA | AJ302735.1 | - | 4 - 5 | 365 | 129 | |
| AJ347709.1 | - | 4 - 5 | 366 | 129 | ||
| AK295971.1 | - | 4 - 5 | 377 | 129 | ||
| BC001257.1 | - | 4 - 5 | 366 | 3454182 | 129 | |
| DA729622.1 | - | 4 - 5 | 399 | 129 | ||
| U73704.1 | - | 4 - 5 | 377 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002866.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
neural tube closure
negative regulation of protein ubiquitination
regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of gene expression, epigenetic
negative regulation of T cell proliferation
positive regulation of phosphorylation
muscle cell differentiation
positive regulation of interleukin-2 biosynthetic process
positive regulation of cytokine secretion
hepatocyte growth factor receptor binding
protein binding
ubiquitin protein ligase binding
ubiquitin-protein transferase inhibitor activity
vasculogenesis
neural tube closure
negative regulation of protein ubiquitination
regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of gene expression, epigenetic
negative regulation of T cell proliferation
positive regulation of phosphorylation
muscle cell differentiation
positive regulation of interleukin-2 biosynthetic process
positive regulation of cytokine secretion
hepatocyte growth factor receptor binding
protein binding
ubiquitin protein ligase binding
ubiquitin-protein transferase inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044696.hg.1 | ENST00000370360.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044696.hg.1 | ENST00000534881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044696.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044696.hg.1 | TC01002866.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100211156.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100211156.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100211156.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100211156.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100211156.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100211156.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100211156.hg.1 | TC0100014921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |

GLMN
Genomic Map