Assay Details
| Gene Symbol: RGS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033345.3 | NP_203131.1 | 2 - 3 | 187 | 66 | |
| XM_011510089.2 | XP_011508391.1 | 3 - 4 | 1227 | 66 | ||
| XM_011510091.2 | XP_011508393.1 | 2 - 3 | 204 | 66 | ||
| XM_017002631.1 | XP_016858120.1 | 6 - 7 | 677 | 66 | ||
| XM_017002632.1 | XP_016858121.1 | 4 - 5 | 508 | 66 | ||
| XM_017002633.1 | XP_016858122.1 | 3 - 4 | 706 | 66 | ||
| XM_017002634.1 | XP_016858123.1 | 3 - 4 | 385 | 66 | ||
| GenBank mRNA | BC069718.1 | - | 2 - 3 | 137 | 7262591 | 66 |
| BX281622.1 | - | 2 - 3 | 198 | 66 | ||
| BX954018.1 | - | 2 - 3 | 91 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011510091.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | ENST00000570153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000258302.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000367557.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000483095.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000491420.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000508450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000515211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | ENST00000570153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_001102450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016633.hg.1 | NM_033345.3 | Refseq |

RGS8
Genomic Map