Assay Details
| Gene Symbol: GLMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053274.2 | NP_444504.1 | 13 - 14 | 1326 | 131 | |
| XM_005270401.3 | XP_005270458.1 | 12 - 13 | 1188 | 131 | ||
| XM_006710309.2 | XP_006710372.1 | 13 - 14 | 1328 | 131 | ||
| XM_011540546.2 | XP_011538848.1 | 15 - 16 | 1489 | 131 | ||
| XM_017000137.1 | XP_016855626.1 | 13 - 14 | 1694 | 131 | ||
| XM_017000140.1 | XP_016855629.1 | 12 - 13 | 1304 | 131 | ||
| XM_017000144.1 | XP_016855633.1 | 12 - 13 | 1212 | 131 | ||
| GenBank mRNA | AJ302735.1 | - | 13 - 14 | 1292 | 131 | |
| AJ347709.1 | - | 13 - 14 | 1293 | 131 | ||
| BC001257.1 | - | 13 - 14 | 1293 | 3454182 | 131 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002866.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
neural tube closure
negative regulation of protein ubiquitination
regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of gene expression, epigenetic
negative regulation of T cell proliferation
positive regulation of phosphorylation
muscle cell differentiation
positive regulation of interleukin-2 biosynthetic process
positive regulation of cytokine secretion
hepatocyte growth factor receptor binding
protein binding
ubiquitin protein ligase binding
ubiquitin-protein transferase inhibitor activity
vasculogenesis
neural tube closure
negative regulation of protein ubiquitination
regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of gene expression, epigenetic
negative regulation of T cell proliferation
positive regulation of phosphorylation
muscle cell differentiation
positive regulation of interleukin-2 biosynthetic process
positive regulation of cytokine secretion
hepatocyte growth factor receptor binding
protein binding
ubiquitin protein ligase binding
ubiquitin-protein transferase inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01044685.hg.1 | ENST00000370360.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044685.hg.1 | ENST00000495852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044685.hg.1 | ENST00000534881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044685.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044685.hg.1 | TC01002866.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000463560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000471465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000487911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014921.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100211139.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100211139.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100211139.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100211139.hg.1 | ENST00000370360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100211139.hg.1 | ENST00000495852.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100211139.hg.1 | NM_053274.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100211139.hg.1 | TC0100014921.hg.1 | Clariom D TC |

GLMN
Genomic Map