Assay Details
| Gene Symbol: PDIA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004911.4 | NP_004902.1 | 2 - 3 | 557 | 111 | |
| XM_006716185.2 | XP_006716248.1 | 2 - 3 | 557 | 111 | ||
| GenBank mRNA | AK290971.1 | - | 2 - 3 | 342 | 111 | |
| AK313299.1 | - | 2 - 3 | 529 | 111 | ||
| BC000425.2 | - | 2 - 3 | 319 | 2819726 | 111 | |
| BC001928.1 | - | 2 - 3 | 319 | 2819726 | 111 | |
| BC006344.2 | - | 2 - 3 | 507 | 4098367 | 111 | |
| BC011754.2 | - | 2 - 3 | 508 | 3532913 | 111 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001995.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000466592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000466592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | ENST00000466592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030668.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030668.hg.1 | TC07001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030660.hg.1 | ENST00000286091.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030660.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030660.hg.1 | TC07001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030661.hg.1 | ENST00000286091.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030661.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030661.hg.1 | TC07001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030666.hg.1 | ENST00000286091.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030666.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030666.hg.1 | TC07001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030662.hg.1 | ENST00000286091.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030662.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030662.hg.1 | TC07001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030664.hg.1 | ENST00000286091.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030664.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030664.hg.1 | TC07001995.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700193510.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193510.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193510.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193510.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193510.hg.1 | TC0700012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700193512.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193512.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193512.hg.1 | TC0700012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700193504.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193504.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193504.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193504.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193504.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193504.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193504.hg.1 | TC0700012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700193508.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193508.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193508.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193508.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193508.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193508.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193508.hg.1 | TC0700012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700193503.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193503.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193503.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193503.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193503.hg.1 | TC0700012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700193507.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193507.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193507.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193507.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193507.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193507.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193507.hg.1 | TC0700012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700193505.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193505.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193505.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193505.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193505.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700193505.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700193505.hg.1 | TC0700012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000286091.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000413966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000466592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000466592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | ENST00000466592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012968.hg.1 | NM_004911.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 184373 | - | - |

PDIA4
Genomic Map