Assay Details
| Gene Symbol: DYRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003582.2 | NP_003573.2 | 1 - 2 | 242 | 147 | |
| GenBank mRNA | AF186773.1 | - | 1 - 2 | 219 | 147 | |
| AK308082.1 | - | 1 - 2 | 242 | 147 | ||
| AK313245.1 | - | 1 - 2 | 242 | 147 | ||
| AY590695.1 | - | 1 - 2 | 241 | 147 | ||
| Y12735.2 | - | 1 - 2 | 222 | 147 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001725.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313245.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026920.hg.1 | ENST00000367108.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026920.hg.1 | ENST00000367109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026920.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026920.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026920.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026920.hg.1 | TC01001725.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026926.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026926.hg.1 | ENST00000367108.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026926.hg.1 | ENST00000367109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026926.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026926.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026926.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026926.hg.1 | TC01001725.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026919.hg.1 | ENST00000367108.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026919.hg.1 | ENST00000367109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026919.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026919.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026919.hg.1 | TC01001725.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026918.hg.1 | ENST00000367109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026918.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026918.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026918.hg.1 | TC01001725.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100183671.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183671.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183671.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183671.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183671.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183671.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183671.hg.1 | TC0100011382.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183674.hg.1 | TC0100011382.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183672.hg.1 | TC0100011382.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011382.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000367106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000367108.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000367109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000441486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | ENST00000489878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | NM_001004023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | NM_003582.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183681.hg.1 | TC0100011382.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 188708 | - | - |

DYRK3
Genomic Map