Assay Details
| Gene Symbol: TRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317804.1 | NP_001304733.1 | 8 - 9 | 816 | 111 | |
| NM_001317805.1 | NP_001304734.1 | 7 - 8 | 1331 | 111 | ||
| NM_014294.5 | NP_055109.1 | 7 - 8 | 899 | 111 | ||
| XM_017013266.1 | XP_016868755.1 | 6 - 7 | 771 | 111 | ||
| XM_017013267.1 | XP_016868756.1 | 6 - 7 | 1115 | 111 | ||
| GenBank mRNA | AK292777.1 | - | 7 - 8 | 683 | 111 | |
| AK303411.1 | - | 8 - 9 | 744 | 111 | ||
| BC000687.2 | - | 7 - 8 | 721 | 3347823 | 111 | |
| BC020283.1 | - | 7 - 8 | 714 | 111 | ||
| BC037738.1 | - | 7 - 8 | 811 | 5261490 | 111 | |
| BT007359.1 | - | 7 - 8 | 639 | 111 | ||
| CR541654.1 | - | 7 - 8 | 639 | 111 | ||
| CR541739.1 | - | 7 - 8 | 639 | 111 | ||
| X63679.1 | - | 7 - 8 | 760 | 111 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001312.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08016898.hg.1 | ENST00000262213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016898.hg.1 | ENST00000521049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016898.hg.1 | ENST00000521425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016898.hg.1 | ENST00000536748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016898.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016898.hg.1 | TC08001312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016901.hg.1 | ENST00000262213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016901.hg.1 | ENST00000521425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016901.hg.1 | ENST00000536748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016901.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016901.hg.1 | TC08001312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016897.hg.1 | ENST00000262213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016897.hg.1 | ENST00000521049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016897.hg.1 | ENST00000521425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016897.hg.1 | ENST00000536748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016897.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016897.hg.1 | TC08001312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000518678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010765.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173062.hg.1 | TC0800010765.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | ENST00000520700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173067.hg.1 | TC0800010765.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | ENST00000262213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | ENST00000521049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | ENST00000521425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | NM_014294.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173060.hg.1 | TC0800010765.hg.1 | Clariom D TC |

TRAM1
Genomic Map