Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ras related dexamethasone induced 1 | RASD1 |
AGS1, DEXRAS1, MGC:26290 |
-54.0kb | Sense | Chr. 17 - 17494437 - 17496395 on NCBI Build GRCh38 |
| Downstream |
Smith-Magenis syndrome chromosome region, candidate 2 (non-protein coding) |
SMCR2 | TCONS_00025215 | 122.9kb | Sense | Chr. 17 - 17674026 - 17677688 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map