Assay Details
| Gene Symbol: SLC30A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018713.2 | NP_061183.2 | 1 - 2 | 853 | 77 | |
| GenBank mRNA | AY212919.1 | - | 1 - 2 | 804 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003832.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058527.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058527.hg.1 | ENST00000536992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058527.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058527.hg.1 | TC01003832.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058524.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058524.hg.1 | ENST00000536446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058524.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058524.hg.1 | TC01003832.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058528.hg.1 | ENST00000536992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058528.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058528.hg.1 | TC01003832.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231343.hg.1 | TC0100017358.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231340.hg.1 | TC0100017358.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | ENST00000484239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017358.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | ENST00000366926.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231342.hg.1 | TC0100017358.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100231344.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231344.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231344.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231344.hg.1 | ENST00000356609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231344.hg.1 | NM_018713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231344.hg.1 | NR_046437.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231344.hg.1 | TC0100017358.hg.1 | Clariom D TC |

SLC30A10
Genomic Map