| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009220.hg.1 |
ENST00000582558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009220.hg.1 |
ENST00000582558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11015612.hg.1 |
ENST00000500108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11015612.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11015612.hg.1 |
TC11001240.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11015611.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11015611.hg.1 |
TC11001240.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006448.hg.1 |
ENST00000508004.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006448.hg.1 |
ENST00000508004.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006448.hg.1 |
ENST00000602429.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006448.hg.1 |
ENST00000602429.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006448.hg.1 |
ENST00000602429.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006448.hg.1 |
ENST00000602756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006448.hg.1 |
ENST00000602756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006448.hg.1 |
ENST00000602756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11015610.hg.1 |
ENST00000500108.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11015610.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11015610.hg.1 |
TC11001240.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100172931.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100172931.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100172931.hg.1 |
TC1100009681.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100172927.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100172927.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100172927.hg.1 |
TC1100009681.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700009220.hg.1 |
ENST00000582558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009220.hg.1 |
ENST00000582558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009220.hg.1 |
ENST00000584705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006448.hg.1 |
ENST00000508004.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006448.hg.1 |
ENST00000508004.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006448.hg.1 |
ENST00000602429.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006448.hg.1 |
ENST00000602429.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006448.hg.1 |
ENST00000602429.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006448.hg.1 |
ENST00000602756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006448.hg.1 |
ENST00000602756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006448.hg.1 |
ENST00000602756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100172929.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100172929.hg.1 |
NR_026967.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100172929.hg.1 |
TC1100009681.hg.1 |
Clariom D TC |