| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003667.hg.1 |
ENST00000311856.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003667.hg.1 |
ENST00000450935.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003667.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003667.hg.1 |
TC03000189.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149477.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149477.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149477.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149477.hg.1 |
ENST00000311856.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149477.hg.1 |
ENST00000415844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149477.hg.1 |
NM_004256.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149477.hg.1 |
TC0300007073.hg.1 |
Clariom D TC |