Assay Details
| Gene Symbol: ZER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006336.3 | NP_006327.2 | 1 - 2 | 357 | 102 | |
| XM_005251646.2 | XP_005251703.1 | 1 - 2 | 352 | 102 | ||
| XM_017014189.1 | XP_016869678.1 | 1 - 2 | 352 | 102 | ||
| GenBank mRNA | AK314911.1 | - | 1 - 2 | 105 | 102 | |
| BC033884.1 | - | 1 - 2 | 326 | 102 | ||
| BC052563.1 | - | 1 - 2 | 342 | 6303965 | 102 | |
| HY170548.1 | - | 1 - 2 | 357 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001638.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK314911.1,BC033884.1,BC052563.1,NM_006336.3,XM_005251646.2,XM_017014189.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09020800.hg.1 | ENST00000291900.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020800.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020800.hg.1 | TC09001638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020802.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020802.hg.1 | TC09001638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020798.hg.1 | ENST00000291900.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020798.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020798.hg.1 | TC09001638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020799.hg.1 | ENST00000291900.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020799.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020799.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020799.hg.1 | TC09001638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020801.hg.1 | ENST00000291900.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020801.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020801.hg.1 | TC09001638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179743.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179743.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179743.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179743.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179743.hg.1 | TC0900011655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179738.hg.1 | TC0900011655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900179741.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179741.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179741.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179741.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179741.hg.1 | TC0900011655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900179742.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179742.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179742.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179742.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179742.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179742.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179742.hg.1 | TC0900011655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011655.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | ENST00000414921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | ENST00000427848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179739.hg.1 | TC0900011655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900179740.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179740.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179740.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179740.hg.1 | ENST00000291900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179740.hg.1 | ENST00000494461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179740.hg.1 | NM_006336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179740.hg.1 | TC0900011655.hg.1 | Clariom D TC |

ZER1
Genomic Map