Assay Details
| Gene Symbol: LNP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085451.1 | NP_001078920.1 | 2 - 3 | 1435 | 75 | |
| GenBank mRNA | AK056320.1 | - | 2 - 3 | 1081 | 75 | |
| BC040562.1 | - | 2 - 3 | 1437 | 75 | ||
| BC126362.1 | - | 2 - 3 | 825 | 8992078 | 75 | |
| BC130480.1 | - | 2 - 3 | 825 | 40146511 | 75 | |
| BC144031.1 | - | 2 - 3 | 825 | 9052551 | 75 | |
More Information
Set Membership: |
|

LNP1
Genomic Map