Assay Details
| Gene Symbol: LRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004525.2 | NP_004516.2 | 43 - 44 | 8425 | 84 | |
| XM_011511183.2 | XP_011509485.1 | 43 - 44 | 8426 | 84 | ||
| XM_011511184.2 | XP_011509486.1 | 28 - 29 | 5938 | 84 | ||
| GenBank mRNA | AY265357.1 | - | 43 - 44 | 8279 | 84 | |
| AY265358.1 | - | 43 - 44 | 8276 | 84 | ||
| U04441.1 | - | 1 - 2 | 100 | 84 | ||
| U33837.1 | - | 43 - 44 | 8316 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002509.hg.1
Set Membership: |
|

LRP2
Genomic Map