Assay Details
| Gene Symbol: GIGYF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022574.4 | NP_072096.2 | 12 - 13 | 2472 | 53 | |
| XM_005250532.1 | XP_005250589.1 | 14 - 15 | 2253 | 53 | ||
| XM_011516472.1 | XP_011514774.1 | 14 - 15 | 2253 | 53 | ||
| XM_011516477.1 | XP_011514779.1 | 14 - 15 | 2253 | 53 | ||
| XM_017012526.1 | XP_016868015.1 | 14 - 15 | 2253 | 53 | ||
| XM_017012527.1 | XP_016868016.1 | 16 - 17 | 2151 | 53 | ||
| XM_017012528.1 | XP_016868017.1 | 15 - 16 | 1763 | 53 | ||
| XM_017012529.1 | XP_016868018.1 | 15 - 16 | 1770 | 53 | ||
| XM_017012530.1 | XP_016868019.1 | 15 - 16 | 2024 | 53 | ||
| XM_017012531.1 | XP_016868020.1 | 10 - 11 | 1276 | 53 | ||
| GenBank mRNA | AY176044.1 | - | 12 - 13 | 2673 | 53 | |
| BC129991.1 | - | 6 - 7 | 620 | 53 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001676.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07025888.hg.1 | ENST00000275732.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025888.hg.1 | ENST00000539430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025888.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025888.hg.1 | TC07001676.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186331.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186331.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186331.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186331.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186331.hg.1 | TC0700012013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000464111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000471340.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012013.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186330.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186330.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186330.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186330.hg.1 | ENST00000275732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186330.hg.1 | ENST00000472105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700186330.hg.1 | NM_022574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700186330.hg.1 | TC0700012013.hg.1 | Clariom D TC |

GIGYF1
Genomic Map