| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039786.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039786.hg.1 |
ENST00000372684.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039786.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039786.hg.1 |
TC01002558.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013837.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100203496.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100203496.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100203496.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100203496.hg.1 |
ENST00000372683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100203496.hg.1 |
ENST00000372684.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100203496.hg.1 |
NM_014747.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100203496.hg.1 |
TC0100013837.hg.1 |
Clariom D TC |