Assay Details
| Gene Symbol: MPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005373.2 | NP_005364.1 | 3 - 4 | 439 | 86 | |
| XM_017001320.1 | XP_016856809.1 | 2 - 3 | 1539 | 86 | ||
| GenBank mRNA | DQ234353.1 | - | 3 - 4 | 373 | 86 | |
| M90102.1 | - | 3 - 4 | 394 | 86 | ||
| M90103.1 | - | 3 - 4 | 394 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000544.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01008640.hg.1 | ENST00000372468.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008640.hg.1 | ENST00000372470.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008640.hg.1 | ENST00000413998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008640.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008640.hg.1 | TC01000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008056.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | ENST00000372470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | ENST00000413998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | ENST00000612993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | NM_005373.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156871.hg.1 | TC0100008056.hg.1 | Clariom D TC |

MPL
Genomic Map