Assay Details
| Gene Symbol: RCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136204.2 | NP_001129676.1 | 1 - 2 | 328 | 72 | |
| NM_018715.3 | NP_061185.1 | 2 - 3 | 497 | 72 | ||
| GenBank mRNA | AB040903.1 | - | 2 - 3 | 428 | 72 | |
| AJ421269.1 | - | 2 - 3 | 514 | 72 | ||
| BC042141.1 | - | 1 - 2 | 293 | 5932549 | 72 | |
| BC053908.1 | - | 1 - 2 | 326 | 6502476 | 72 | |
| BC142946.1 | - | 1 - 2 | 84 | 72 | ||
| CR991144.1 | - | 1 - 2 | 328 | 72 | ||
| DR000303.1 | - | 2 - 3 | 503 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002289.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
sister chromatid cohesion
mitotic nuclear division
integrin-mediated signaling pathway
regulation of fibroblast migration
positive regulation of G2/M transition of mitotic cell cycle
regulation of cell migration
negative regulation of GTPase activity
establishment of protein localization
focal adhesion assembly
cell division
negative regulation of focal adhesion assembly
positive regulation of attachment of spindle microtubules to kinetochore
chromosome passenger complex localization to kinetochore
activation of GTPase activity
negative regulation of substrate adhesion-dependent cell spreading
regulation of ruffle assembly
protein binding
microtubule binding
protein kinase binding
protein domain specific binding
small GTPase binding
poly(A) RNA binding
Rac GTPase binding
sister chromatid cohesion
mitotic nuclear division
integrin-mediated signaling pathway
regulation of fibroblast migration
positive regulation of G2/M transition of mitotic cell cycle
regulation of cell migration
negative regulation of GTPase activity
establishment of protein localization
focal adhesion assembly
cell division
negative regulation of focal adhesion assembly
positive regulation of attachment of spindle microtubules to kinetochore
chromosome passenger complex localization to kinetochore
activation of GTPase activity
negative regulation of substrate adhesion-dependent cell spreading
regulation of ruffle assembly
protein binding
microtubule binding
protein kinase binding
protein domain specific binding
small GTPase binding
poly(A) RNA binding
Rac GTPase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012121.hg.1 | ENST00000405832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012121.hg.1 | ENST00000405832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012121.hg.1 | ENST00000405832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012121.hg.1 | ENST00000578149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034902.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034902.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034902.hg.1 | TC01002289.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008080.hg.1 | ENST00000492126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000474892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000474892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | ENST00000583469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008080.hg.1 | ENST00000492126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012121.hg.1 | ENST00000405832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012121.hg.1 | ENST00000405832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012121.hg.1 | ENST00000405832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012121.hg.1 | ENST00000578149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196731.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196731.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196731.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196731.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196731.hg.1 | TC0100013087.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375433.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000375436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000474892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000474892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | ENST00000583469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_001136204.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013087.hg.1 | NM_018715.3 | Refseq |

RCC2
Genomic Map