Assay Details
| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039999.2 | NP_001035088.2 | 4 - 5 | 980 | 81 | |
| XM_017024953.1 | XP_016880442.1 | 4 - 5 | 1064 | 81 | ||
| GenBank mRNA | BX365956.2 | - | 1 - 2 | 69 | 81 | |
| DN831927.1 | - | 3 - 4 | 361 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001229.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000580115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | ENST00000580115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016616.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016616.hg.1 | ENST00000388995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016616.hg.1 | ENST00000399096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016616.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016616.hg.1 | TC17001229.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016613.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016613.hg.1 | ENST00000388995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016613.hg.1 | ENST00000399096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016613.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016613.hg.1 | TC17001229.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016617.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016617.hg.1 | ENST00000388995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016617.hg.1 | ENST00000399096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016617.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016617.hg.1 | TC17001229.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016618.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016618.hg.1 | ENST00000399096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016618.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016618.hg.1 | TC17001229.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016615.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016615.hg.1 | ENST00000388995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016615.hg.1 | ENST00000399096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016615.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016615.hg.1 | TC17001229.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016620.hg.1 | ENST00000399096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016620.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17016620.hg.1 | TC17001229.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | TC1700009992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178404.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178412.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178412.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178412.hg.1 | TC1700009992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700178412.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178412.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | TC1700009992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178407.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | TC1700009992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178408.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000580115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | ENST00000580115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009992.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | TC1700009992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | ENST00000399096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178406.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178410.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178410.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178410.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178410.hg.1 | TC1700009992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700178410.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178410.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178410.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178409.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178409.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178409.hg.1 | NM_001039999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700178409.hg.1 | TC1700009992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700178409.hg.1 | ENST00000345041.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178409.hg.1 | ENST00000388995.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700178409.hg.1 | NM_001039999.2 | Refseq |

FAM83G
Genomic Map