Assay Details
| Gene Symbol: TSC22D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030935.4 | NP_112197.1 | 1 - 2 | 486 | 74 | |
| GenBank mRNA | AJ133115.1 | - | 1 - 2 | 75 | ||
| BC001486.2 | - | 1 - 2 | 486 | 2966929 | 74 | |
| BC001966.1 | - | 1 - 2 | 339 | 3457297 | 74 | |
| BC002972.1 | - | 1 - 2 | 393 | 3538175 | 74 | |
| BC010406.2 | - | 1 - 2 | 400 | 3832780 | 74 | |
| BC031622.1 | - | 1 - 2 | 376 | 5174360 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003370.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ133115.1,BC001486.2,BC001966.1,BC002972.1,BC010406.2,BC031622.1,NM_030935.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07025703.hg.1 | ENST00000300181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025703.hg.1 | ENST00000393991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025703.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025703.hg.1 | TC07003370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025708.hg.1 | ENST00000300181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025708.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025708.hg.1 | TC07003370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025701.hg.1 | ENST00000300181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025701.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025701.hg.1 | TC07003370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025707.hg.1 | ENST00000300181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025707.hg.1 | ENST00000393991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025707.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025707.hg.1 | TC07003370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025704.hg.1 | ENST00000300181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025704.hg.1 | ENST00000393991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025704.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025704.hg.1 | TC07003370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025702.hg.1 | ENST00000300181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025702.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025702.hg.1 | TC07003370.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201350.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201350.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201350.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201350.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201350.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201350.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201350.hg.1 | TC0700013596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700201344.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201344.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201344.hg.1 | TC0700013596.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 172824 | - | - |

TSC22D4
Genomic Map