Assay Details
| Gene Symbol: CCT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256721.1 | NP_001243650.1 | 4 - 5 | 600 | 99 | |
| NM_006430.3 | NP_006421.2 | 5 - 6 | 690 | 99 | ||
| GenBank mRNA | AF026291.1 | - | 5 - 6 | 628 | 99 | |
| AK129860.1 | - | 5 - 6 | 599 | 99 | ||
| AK290538.1 | - | 5 - 6 | 657 | 99 | ||
| AK294665.1 | - | 4 - 5 | 434 | 99 | ||
| AK295330.1 | - | 5 - 6 | 463 | 99 | ||
| AK303082.1 | - | 4 - 5 | 585 | 99 | ||
| AK307856.1 | - | 6 - 7 | 624 | 99 | ||
| AK308942.1 | - | 5 - 6 | 607 | 99 | ||
| AK312586.1 | - | 5 - 6 | 632 | 99 | ||
| AK315975.1 | - | 6 - 7 | 866 | 99 | ||
| BC106933.1 | - | 5 - 6 | 623 | 40033761 | 99 | |
| BC106934.1 | - | 5 - 6 | 626 | 40033762 | 99 | |
| U38846.1 | - | 5 - 6 | 526 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001895.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
binding of sperm to zona pellucida
positive regulation of telomere maintenance via telomerase
protein stabilization
positive regulation of telomerase activity
scaRNA localization to Cajal body
toxin transport
positive regulation of establishment of protein localization to telomere
positive regulation of protein localization to Cajal body
positive regulation of telomerase RNA localization to Cajal body
protein binding
ATP binding
poly(A) RNA binding
unfolded protein binding
protein folding
binding of sperm to zona pellucida
positive regulation of telomere maintenance via telomerase
protein stabilization
positive regulation of telomerase activity
scaRNA localization to Cajal body
toxin transport
positive regulation of establishment of protein localization to telomere
positive regulation of protein localization to Cajal body
positive regulation of telomerase RNA localization to Cajal body
protein binding
ATP binding
poly(A) RNA binding
unfolded protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000461370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000461370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030391.hg.1 | ENST00000394440.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030391.hg.1 | ENST00000538252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030391.hg.1 | ENST00000544079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030391.hg.1 | ENST00000544185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030391.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030391.hg.1 | TC02001895.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193126.hg.1 | TC0200012755.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000394440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000461370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000461370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000461540.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | ENST00000544079.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_001256721.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012755.hg.1 | NM_006430.3 | Refseq |

CCT4
Genomic Map