Assay Details
| Gene Symbol: PADI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207421.4 | NP_997304.3 | 13 - 14 | 1670 | 109 | |
| GenBank mRNA | AY422079.1 | - | 13 - 14 | 1620 | 109 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000233.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01003742.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003742.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003742.hg.1 | TC01000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003741.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003741.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003741.hg.1 | TC01000233.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007108.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149622.hg.1 | TC0100007108.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | ENST00000358481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | ENST00000434762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | ENST00000619609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | NM_207421.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149623.hg.1 | TC0100007108.hg.1 | Clariom D TC |

PADI6
Genomic Map