Assay Details
| Gene Symbol: TSC22D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130119.1 | 1 - 2 | 485 | 68 | ||
| GenBank mRNA | BX341171.2 | - | 1 - 2 | 437 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201346.hg.1 | TC0700013596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000423266.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000456330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013596.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201351.hg.1 | TC0700013596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201355.hg.1 | TC0700013596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | ENST00000393991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | ENST00000493217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201352.hg.1 | TC0700013596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | ENST00000300181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | NM_001303043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | NM_030935.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | NR_130118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | NR_130119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201356.hg.1 | TC0700013596.hg.1 | Clariom D TC |

TSC22D4
Genomic Map