| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002605.hg.1 |
ENST00000269491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002605.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002605.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002605.hg.1 |
TC18000227.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002604.hg.1 |
ENST00000269491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002604.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002604.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002604.hg.1 |
TC18000227.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
TC1800007546.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151763.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151765.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151765.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151765.hg.1 |
TC1800007546.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800151765.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151765.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
NM_080474.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
TC1800007546.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
ENST00000269491.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
ENST00000382768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
NM_001307928.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151764.hg.1 |
NM_080474.2 |
Refseq |